COMH93D605C5 Combi-Blocks, Inc.

2,5-Pyrrolidinedione, 1-[(3-iodobenzoyl)oxy]-

  • MDL number MFCD27964177
  • Cas Number : 91487-18-2
  • Molecular Weight : 345.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6657

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6657
Availability details for
Quantity
  • Availability
  • Add to Cart