COMH93D60616 Combi-Blocks, Inc.

10-Trifluoroacetamidodec-1-ene

  • MDL number MFCD27930113
  • Cas Number : 85657-91-6
  • Molecular Weight : 251.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6767

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6767
Availability details for
Quantity
  • Availability
  • Add to Cart