COMH93D6061A Combi-Blocks, Inc.

4-Bromo-5-chloro-1-methylpyridin-2(1h)-one

  • MDL number MFCD30179619
  • Cas Number : 1785263-12-8
  • Molecular Weight : 222.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6772

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6772
Availability details for
Quantity
  • Availability
  • Add to Cart