COMH93D60623 Combi-Blocks, Inc.

2-Chloro-n-(4-chlorophenyl)-9-methyl-9h-purin-6-amine

  • MDL number MFCD28142676
  • Cas Number : 1062592-41-9
  • Molecular Weight : 294.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6783

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6783
Availability details for
Quantity
  • Availability
  • Add to Cart