COMH93D6062E Combi-Blocks, Inc.

3,3-Difluoro-1-(2,2,2-trifluoroacetyl)piperidine-2-carboxylic acid

  • MDL number MFCD28343694
  • Cas Number : 1039364-78-7
  • Molecular Weight : 261.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6798

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6798
Availability details for
Quantity
  • Availability
  • Add to Cart