COMH93D6064C Combi-Blocks, Inc.

6-Chloro-3-((2,3-dichloropyridin-4-yl)thio)pyrazin-2-amine

  • MDL number MFCD30530824
  • Cas Number : 1801693-92-4
  • Molecular Weight : 307.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6832

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6832
Availability details for
Quantity
  • Availability
  • Add to Cart