COMH93D60654 Combi-Blocks, Inc.

3-Chloro-5-nitro-1h-pyrazole

  • MDL number MFCD29762425
  • Cas Number : 1369959-12-5
  • Molecular Weight : 147.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6841

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6841
Availability details for
Quantity
  • Availability
  • Add to Cart