COMH93D60681 Combi-Blocks, Inc.

2,4-Dichloropyrido[3',2':4,5]furo[3,2-d]pyrimidine

  • MDL number MFCD22572868
  • Cas Number : 1268241-74-2
  • Molecular Weight : 240.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6898

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6898
Availability details for
Quantity
  • Availability
  • Add to Cart