COMH93D606B4 Combi-Blocks, Inc.

1-Acetyl-2-methyl-2,3-dihydroquinolin-4(1h)-one

  • MDL number MFCD28366233
  • Cas Number : 128649-34-3
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6964

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6964
Availability details for
Quantity
  • Availability
  • Add to Cart