COMH93D606B5 Combi-Blocks, Inc.

3-Bromo-1,7-naphthyridin-8-amine

  • MDL number MFCD30828924
  • Cas Number : 1838674-91-1
  • Molecular Weight : 224.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6965

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6965
Availability details for
Quantity
  • Availability
  • Add to Cart