COMH93D606E7 Combi-Blocks, Inc.

4-(6-Bromoimidazo[1,2-a]pyridin-3-yl)pyrimidin-2-amine

  • MDL number MFCD30185867
  • Cas Number : 453510-84-4
  • Molecular Weight : 290.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7031

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7031
Availability details for
Quantity
  • Availability
  • Add to Cart