COMH93D6088A Combi-Blocks, Inc.

6-Fluoroisoquinolin-4-amine

  • MDL number MFCD28539787
  • Cas Number : 1780542-36-0
  • Molecular Weight : 162.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7576

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7576
Availability details for
Quantity
  • Availability
  • Add to Cart