COMH93D608AF Combi-Blocks, Inc.

(3-(Pyridin-3-yl)phenyl)boronic acid pinacol ester hcl

  • MDL number MFCD31629662
  • Cas Number : 2304635-18-3
  • Molecular Weight : 317.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7616

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7616
Availability details for
Quantity
  • Availability
  • Add to Cart