COMH93D608B0 Combi-Blocks, Inc.

2-Bromo-4-methoxy-6-methylaniline

  • MDL number MFCD28098569
  • Cas Number : 1100394-71-5
  • Molecular Weight : 216.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7617

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7617
Availability details for
Quantity
  • Availability
  • Add to Cart