COMH93D608C4 Combi-Blocks, Inc.

(4-Methoxy-4-oxobutyl) triphenylphosphonium bromide

  • MDL number MFCD28969545
  • Cas Number : 13395-34-1
  • Molecular Weight : 443.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7645

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7645
Availability details for
Quantity
  • Availability
  • Add to Cart