COMH93D608DD Combi-Blocks, Inc.

Trans-hexahydro-cyclopenta[c]pyrrole-2,5-dicarboxylic acid mono-tert-butyl ester

  • MDL number MFCD29638109
  • Molecular Weight : 255.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7677

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7677
Availability details for
Quantity
  • Availability
  • Add to Cart