COMH93D608DF Combi-Blocks, Inc.

[1-(2-Bromo-5-chlorophenyl)ethyl](ethyl)amine

  • MDL number MFCD30224798
  • Cas Number : 1690625-75-2
  • Molecular Weight : 262.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7679

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7679
Availability details for
Quantity
  • Availability
  • Add to Cart