COMH93D60912 Combi-Blocks, Inc.

2-(4-(Trifluoromethyl)phenyl)cyclopropanecarbonitrile

  • MDL number MFCD31651744
  • Molecular Weight : 211.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7738

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7738
Availability details for
Quantity
  • Availability
  • Add to Cart