COMH93D60918 Combi-Blocks, Inc.

2-(But-3-en-1-yloxy)-4-fluoro-1-nitrobenzene

  • MDL number MFCD23725194
  • Cas Number : 154576-90-6
  • Molecular Weight : 211.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7750

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7750
Availability details for
Quantity
  • Availability
  • Add to Cart