COMH93D60980 Combi-Blocks, Inc.

Benzyl (4-methylazepan-4-yl)carbamate hcl

  • MDL number MFCD31651866
  • Cas Number : 2225878-72-6
  • Molecular Weight : 298.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7875

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7875
Availability details for
Quantity
  • Availability
  • Add to Cart