COMH93D60987 Combi-Blocks, Inc.

1-Methyl-4-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1h-pyrazole

  • MDL number MFCD28166950
  • Cas Number : 1534350-44-1
  • Molecular Weight : 284.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7885

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7885
Availability details for
Quantity
  • Availability
  • Add to Cart