COMH93D609AB Combi-Blocks, Inc.

Ethyl 3-bromo-1h-pyrrolo[2,3-b]pyridine-5-carboxylate

  • MDL number MFCD31629639
  • Cas Number : 2090461-66-6
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7929

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7929
Availability details for
Quantity
  • Availability
  • Add to Cart