COMH93D609C4 Combi-Blocks, Inc.

3-((3,4-Dichlorophenoxy)methyl)azetidine

  • MDL number MFCD26400708
  • Cas Number : 1332301-47-9
  • Molecular Weight : 232.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7959

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7959
Availability details for
Quantity
  • Availability
  • Add to Cart