COMH93D609DF Combi-Blocks, Inc.

2-Chloro-5-((4-fluorobenzyl)oxy)pyridine

  • MDL number MFCD23903420
  • Cas Number : 1542425-20-6
  • Molecular Weight : 237.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7993

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-7993
Availability details for
Quantity
  • Availability
  • Add to Cart