COMH93D60A09 Combi-Blocks, Inc.

Cis-3-tosyl-3-azabicyclo[3.2.0]heptan-6-one

  • MDL number MFCD31563653
  • Molecular Weight : 265.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8041

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8041
Availability details for
Quantity
  • Availability
  • Add to Cart