COMH93D60A17 Combi-Blocks, Inc.

3-Amino-n-(propan-2-yl)-4-(trifluoromethoxy)benzamide

  • MDL number MFCD27843047
  • Molecular Weight : 262.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8060

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8060
Availability details for
Quantity
  • Availability
  • Add to Cart