COMH93D60A3E Combi-Blocks, Inc.

(S)-3-Amino-1-(3,5-dimethoxybenzyl) pyrrolidin-2-one hcl

  • MDL number MFCD31560499
  • Cas Number : 2227107-80-2
  • Molecular Weight : 286.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8105

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8105
Availability details for
Quantity
  • Availability
  • Add to Cart