COMH93D60A76 Combi-Blocks, Inc.

tert-Butyl 2-(1-amino-2-methylpropan-2-yl)hydrazinecarboxylate

  • MDL number MFCD24645034
  • Cas Number : 1354825-86-7
  • Molecular Weight : 203.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8174

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8174
Availability details for
Quantity
  • Availability
  • Add to Cart