COMH93D60A89 Combi-Blocks, Inc.

5-Hydroxytetrahydro-2h-pyran-2-one

  • MDL number MFCD26620324
  • Cas Number : 33691-73-5
  • Molecular Weight : 116.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8194

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8194
Availability details for
Quantity
  • Availability
  • Add to Cart