COMH93D60A8D Combi-Blocks, Inc.

Fmoc-(r)-3-amino-4-(4-methoxy-phenyl)-butyric acid

  • MDL number MFCD22887415
  • Cas Number : 1421258-65-2
  • Molecular Weight : 431.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8202

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8202
Availability details for
Quantity
  • Availability
  • Add to Cart