COMH93D60A98 Combi-Blocks, Inc.

2-Bromo-7-(trifluoromethyl)-[1,2,4]triazolo[1,5-a]pyridine

  • MDL number MFCD29041513
  • Cas Number : 1397287-53-4
  • Molecular Weight : 266.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8216

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8216
Availability details for
Quantity
  • Availability
  • Add to Cart