COMH93D60AAA Combi-Blocks, Inc.

2-(2H-1,2,3-Triazol-2-yl)ethan-1-amine hydrochloride

  • MDL number MFCD28125978
  • Cas Number : 4320-99-4
  • Molecular Weight : 148.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8236

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8236
Availability details for
Quantity
  • Availability
  • Add to Cart