COMH93D60AD2 Combi-Blocks, Inc.

tert-Butyl 3-(4-amino-3-methoxyphenoxy)azetidine-1-carboxylate

  • MDL number MFCD28963099
  • Cas Number : 1224709-37-8
  • Molecular Weight : 294.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8290

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8290
Availability details for
Quantity
  • Availability
  • Add to Cart