COMH93D60B0C Combi-Blocks, Inc.

4-(Difluoromethyl)pyrimidin-2(1h)-one

  • MDL number MFCD29921222
  • Cas Number : 1522243-39-5
  • Molecular Weight : 146.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8361

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8361
Availability details for
Quantity
  • Availability
  • Add to Cart