COMH93D60B92 Combi-Blocks, Inc.

Methyl 4-(tetrahydro-2h-pyran-2-yloxy)but-2-ynoate

  • MDL number MFCD28046354
  • Cas Number : 69511-49-5
  • Molecular Weight : 198.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8533

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8533
Availability details for
Quantity
  • Availability
  • Add to Cart