COMH93D60BA0 Combi-Blocks, Inc.

1-Benzhydryl-3-chloro-3-methylazetidine

  • MDL number MFCD30490536
  • Cas Number : 1613721-74-6
  • Molecular Weight : 271.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8548

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8548
Availability details for
Quantity
  • Availability
  • Add to Cart