COMH93D60C43 Combi-Blocks, Inc.

Di-tert-butyl 4-(tert-butoxycarbonylamino)-1h-imidazole-1,3(2h)-dicarboxylate

  • MDL number MFCD30730189
  • Molecular Weight : 385.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8761

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8761
Availability details for
Quantity
  • Availability
  • Add to Cart