COMH93D60C86 Combi-Blocks, Inc.

2-Chloro-4-methoxybenzo[d]oxazole

  • MDL number MFCD28803386
  • Cas Number : 1599561-87-1
  • Molecular Weight : 183.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8839

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8839
Availability details for
Quantity
  • Availability
  • Add to Cart