COMH93D60CC8 Combi-Blocks, Inc.

Ethyl 5-formyl-4-nitro-1h-pyrrole-2-carboxylate

  • MDL number MFCD25962723
  • Cas Number : 36131-47-2
  • Molecular Weight : 212.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8925

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8925
Availability details for
Quantity
  • Availability
  • Add to Cart