COMH93D60CCD Combi-Blocks, Inc.

Ethyl 6-(4-bromo-3-formylphenoxy)-2-ethoxynicotinate

  • MDL number MFCD30185084
  • Cas Number : 1187189-41-8
  • Molecular Weight : 394.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8930

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8930
Availability details for
Quantity
  • Availability
  • Add to Cart