COMH93D60CE0 Combi-Blocks, Inc.

3-Amino-n-(cyclopropylmethyl)-5-fluorobenzamide

  • MDL number MFCD24125117
  • Molecular Weight : 208.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8957

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8957
Availability details for
Quantity
  • Availability
  • Add to Cart