COMH93D60CE7 Combi-Blocks, Inc.

(1R,2S,4S)-Rel-7-azabicyclo[2.2.1]heptan-2-ol

  • MDL number MFCD28515427
  • Cas Number : 191468-74-3
  • Molecular Weight : 113.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8968

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-8968
Availability details for
Quantity
  • Availability
  • Add to Cart