COMH93D60D0D Combi-Blocks, Inc.

3-Iodo-5-nitro-1-trityl-1h-indazole

  • MDL number MFCD30471950
  • Cas Number : 473416-34-1
  • Molecular Weight : 531.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9022

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9022
Availability details for
Quantity
  • Availability
  • Add to Cart