COMH93D60D3F Combi-Blocks, Inc.

Chembopharma kb-271109

  • MDL number MFCD28160365
  • Molecular Weight : 372.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9078

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9078
Availability details for
Quantity
  • Availability
  • Add to Cart