COMH93D60D7D Combi-Blocks, Inc.

6-Chloro-4-nitro-3-(trifluoromethyl)-1-((2-(trimethylsilyl)ethoxy)methyl)-1h-pyrrolo[2,3-b]pyridine

  • MDL number MFCD26960978
  • Cas Number : 892414-51-6
  • Molecular Weight : 395.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9157

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9157
Availability details for
Quantity
  • Availability
  • Add to Cart