COMH93D60D8B Combi-Blocks, Inc.

7-Bromo-2-chloro-1,5-naphthyridine

  • MDL number MFCD24848136
  • Cas Number : 1309774-03-5
  • Molecular Weight : 243.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9177

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9177
Availability details for
Quantity
  • Availability
  • Add to Cart