COMH93D60DCE Combi-Blocks, Inc.

1-(5-Bromo-2-fluoro-benzyl)-4-isopropoxy-1h-pyrazole

  • MDL number MFCD31654522
  • Cas Number : 2270910-71-7
  • Molecular Weight : 313.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9258

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9258
Availability details for
Quantity
  • Availability
  • Add to Cart