COMH93D60DF3 Combi-Blocks, Inc.

4-((tert-Butyldimethylsilyl)oxy)-n-(4-chlorophenyl)piperidine-1-carbothioamide

  • MDL number MFCD29917095
  • Cas Number : 1956365-05-1
  • Molecular Weight : 385.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9302

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9302
Availability details for
Quantity
  • Availability
  • Add to Cart