COMH93D60E05 Combi-Blocks, Inc.

Ethyl 3,3-dimethyl-2-phenyloxirane-2-carboxylate

  • MDL number MFCD28978611
  • Cas Number : 59070-02-9
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9326

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9326
Availability details for
Quantity
  • Availability
  • Add to Cart