COMH93D60E17 Combi-Blocks, Inc.

4-Ethylthiazol-5-amine hcl

  • MDL number MFCD31652501
  • Cas Number : 2206568-02-5
  • Molecular Weight : 164.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9345

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-9345
Availability details for
Quantity
  • Availability
  • Add to Cart